missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Optical Adhesive Covers GPLE

Catalog No. A49767
Change view
Click to view available options
Quantity:
25 covers
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A49767 25 covers
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A49767 Supplier Applied Biosystems™ Supplier No. A49767

The Applied Biosystems Optical Adhesive Cover GPLE is a pressure sensitive film that reduces well-to-well contamination and prevents sample evaporation when applied to a microplate. It is optically clear for use with real-time PCR applications.

  • A pressure sensitive film that reduces well-to-well contamination and prevents sample evaporation when applied to a microplate
  • It is optically clear for use with real-time PCR applications
  • Each lot is manufactured in an ISO registered facility, making them ideal for use with diagnostic procedures
  • Covers are easy to use, simply peel the backing away from the adhesive film and place on the microplate
  • Use in combination with the MicroAmp Adhesive Film Applicator to apply pressure and form a tight seal between the microplate and the adhesive film
  • This will reduce the possibility of cross-contamination between sample wells and prevent sample evaporation for consistent real-time PCR data

Specifications

Certifications/Compliance GPLE
Sterility Non-sterile
Includes 25 covers
Quantity 25 covers
For Use With (Application) PCR, qPCR
Content And Storage Store at room temperature and avoid direct exposure to sunlight.
Shipping Condition Room temperature
Product Type Film, Seal, or Applicator
Adhesive Pressure Sensitive
Pierceable No
Plate Compatibility 96/384-well Plate
Show More Show Less

For Laboratory Use