missing translation for 'onlineSavingsMsg'
Learn More

Ophthalmic acid , ≥97.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014314
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014314 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014314 Supplier Merck Emd Millipore Supplier No. 9135110MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

MDL Number MFCD00076874
Synonym H-gamma-Glu-Abu-Gly-OH; L-gamma-Glutamyl-(2 S)-2-aminobutanoylglycine

Specifications

CAS 495-27-2
Linear Formula C11H19N3O6
Quantity 10 mg
Formula Weight 289.29
Percent Purity ≥97.0% (HPLC)