Learn More
Humboldt Riffle-type Sample Splitter with Removable Hopper
Riffle-type sample splitter with removable hopper can be used to divide or halve dry materials such as cement, gravel, powdered ores, coal, coke, sand, soils, etc.
Supplier: Humboldt H 3980
Description
Material poured into the hopper is divided into two equal portions by a series of chutes that discharge the material alternately in opposite directions into separate pans. Splitter is stainless steel construction with cold-rolled steel pans.
All splitters include 2 pans, scoop and cleaning brush.
Identifiants chimiques
| Riffle Sample Splitter | |
| 2 Pans, Scoop, Cleaning Brush | |
| ASTM B215, C136, C702, C778, D421, D424, D457, D806, AASHTO T27, T144, T248 |
| Stainless Steel | |
| Riffle | |
| Divide or halve dry materials such as cement, gravel, powdered ores, coal, coke, sand, soils |
Spécifications
| Riffle Sample Splitter | |
| 2 Pans, Scoop, Cleaning Brush | |
| 12 | |
| Divide or halve dry materials such as cement, gravel, powdered ores, coal, coke, sand, soils | |
| 11.43 cm | |
| 8 in. | |
| 20.32 cm | |
| 11 in. | |
| 1.27 cm | |
| 13.23 cm |
| Stainless Steel | |
| Riffle | |
| ASTM B215, C136, C702, C778, D421, D424, D457, D806, AASHTO T27, T144, T248 | |
| 4.5 in. | |
| 8.5 in. | |
| 21.56 cm | |
| 0.5 in. | |
| 5.2 in. | |
| 27.98 cm |