missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals ONO-RS-082, 98%

Catalog No. AAJ67444FPL
Change view
Click to view available options
Quantity:
20 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67444FPL 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ67444FPL Supplier Thermo Scientific Chemicals Supplier No. J67444FPL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 99754-06-0
Molecular Formula C21H22ClNO3
Synonym 4-Chloro-2-(4-n-pentylcinnamoylamino)benzoic acid;

Specifications

Quantity 20 mg
Description Off-white to yellow
Formula Weight 371.86
Sensitivity Light Sensitive
Melting Point 175°C to 177°C
Product Type ONO-RS-082, 98%
Form Solid
Assay 98%

RUO – Research Use Only