Learn More
New Pig Only Spill Kit in High - Visibility Bag
PIG Oil Absorbents in an easy to see, fast to find, High-Visibility bag. Durable, water-resistant bag with hook-and-loop closure keeps contents clean and dry
Supplier:  New Pig KIT420
Catalog No. 17987835
Only null left
Description
Description
  • DURABLE, WATER-RESISTANT : Bag with hook-and-loop closure keeps contents clean and dry
  • PIG OIL-ONLY SOCKS : Stop spreading spills, Pig Stat-Mat pads absorb quickly
  • ABSORBENTS MEET NFPA 99Standards for static decay to allow use with fuels and flammables
  • TEMPORARY DISPOSAL BAGS Helps make cleanup easier
  • EASY TO LOCATE : High-visibility bag contains PIG Oil Absorbents proven for rapid response
  • Strap for easy carrying and hanging
  • COMPACT AND EASY TO STORE - makes a great truck spill kit
Product Suggestions
Product Suggestions