Learn More
MP Biomedicals™ OneStep Western Blocker and Signal Enhancer (Protein-Free)
Catalog No. MP08X100002
Change view
Click to view available options
Quantity:
500 mL
Catalog No. Quantity
MP08X100002 500 mL
1 options
Catalog No. MP08X100002 Supplier MP Biomedicals™ Supplier No. 08X100002
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
OneStep Western Blocker and Signal Enhancer (Protein-Free) provides blocking, primary and secondary antibody hybridization in one step. This enhances the signal developed with HRP or alkaline phosphatase substrates.
  • This OneStep Blocker buffer not only provides blocking and primary and secondary antibody hybridization in one step but also enhances the signal developed with HRP (horseradish peroxidase) or AP (alkaline phosphatase) substrates.
  • With the three-in-one step procedure, OneStep Blocker is a time and labor economic solution for the time-consuming and laborious Western procedure.
Specifications
Quantity 500 mL
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.