missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ OneFeed™ Supplement
A versatile medium supplement.
$789.86 - $2820.95
Specifications
| Content And Storage | Storage conditions: 2°C to 8°C. Store in cool, dry conditions in well-sealed containers. |
|---|---|
| Cell Line | CHO DG44, BHK-21, CHO-K1, CHO |
| Classification | Animal Origin-free, Chemically-defined, Protein-free |
| Form | Powder |
| Product Type | Media Supplement |
Description
Gibco™ OneFeed™ Supplement is a chemically-defined, animal-free, protein-free feed shown to enhance protein production when used in a variety of commercially available medium.Specifications
| Storage conditions: 2°C to 8°C. Store in cool, dry conditions in well-sealed containers. | |
| Animal Origin-free, Chemically-defined, Protein-free | |
| Media Supplement |
| CHO DG44, BHK-21, CHO-K1, CHO | |
| Powder | |
| Animal origin-free |