missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ OneFeed™ Supplement
GREENER_CHOICE

Catalog No. 670109
Change view
Click to view available options
Quantity:
10 L
50 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
670109 10 L
670108 50 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 670109 Supplier Gibco™ Supplier No. 670109

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A versatile medium supplement.

Gibco™ OneFeed™ Supplement is a chemically-defined, animal-free, protein-free feed shown to enhance protein production when used in a variety of commercially available medium.

Specifications

Content And Storage Storage conditions: 2°C to 8°C. Store in cool, dry conditions in well-sealed containers.
Cell Line CHO DG44, BHK-21, CHO-K1, CHO
Classification Animal Origin-free, Chemically-defined, Protein-free
Form Powder
Product Type Media Supplement
Format Container
Quantity 10 L
Source Animal origin-free