missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Oncomodulin Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5149897
Description
Oncomodulin Polyclonal Antibody for IHC (P), Western Blot
Oncomodulin, encoded by the OCM gene located on chromosome 7p22.3, is a protein that belongs to the parvalbumin family of calcium-binding proteins. It is most prominently found in the inner ear, as well as in certain retinal cells and regenerating neurons. Oncomodulin plays a crucial role in calcium ion homeostasis, which is vital for various cellular processes, including signal transduction, cellular motility, and growth. Notably, oncomodulin has been studied for its role in promoting nerve regeneration, particularly within the central nervous system, where it enhances axonal growth and neuronal repair. This makes it a molecule of significant interest for therapeutic strategies aiming to treat nerve injuries or neurodegenerative diseases. Beyond its neuroregenerative potential, oncomodulin is also expressed in some tumor types, which initially led to its association with cancer-related studies. Current research continues to explore its dual roles in both oncology and neurobiology to better understand its functional implications across different tissues.Identifiants chimiques
| Oncomodulin | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Mouse | |
| Ocm | |
| hCG18255; Ocm; OCM1; Ocm2; OCMN; OM; ONCM; oncmodulin; oncomodulin; oncomodulin 1; oncomodulin 2; oncomodulin-1; Parvalbumin beta | |
| Ocm | |
| Primary | |
| Antigen affinity chromatography, Protein A |
| 0.5 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P51879 | |
| 18261 | |
| Recombinant Oncomodulin (OCM) (Met1-Ser109). | |
| Antibody |
Spécifications
| Oncomodulin | |
| Polyclonal | |
| Unconjugated | |
| Ocm | |
| hCG18255; Ocm; OCM1; Ocm2; OCMN; OM; ONCM; oncmodulin; oncomodulin; oncomodulin 1; oncomodulin 2; oncomodulin-1; Parvalbumin beta | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 18261 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P51879 | |
| Ocm | |
| Recombinant Oncomodulin (OCM) (Met1-Ser109). | |
| 200 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |