missing translation for 'onlineSavingsMsg'
Learn More

Ion Torrent™ Oncomine™ Myeloid Assay GX v2

Numéro de catalogue. A50694
Modifier l'affichage
Click to view available options
Quantité:
32 samples
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
A50694 32 samples
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. A50694 Fournisseur Ion Torrent™ Code fournisseur A50694

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

A comprehensive targeted next-generation sequencing (NGS) assay designed for sensitive detection of myeloid disorder-associated DNA mutations and RNA fusion transcripts in blood and bone marrow samples.

  • A comprehensive targeted next-generation sequencing (NGS) assay
  • Designed for sensitive detection of myeloid disorder-associated DNA mutations and RNA fusion transcripts in blood and bone marrow samples
  • Assay is compatible with the Genexus Integrated Sequencer (A45727) which performs library preparation, sequencing, analysis, and reporting in an automated sample-to-result workflow
  • Kit includes three pools of Ion AmpliSeq oligonucleotide primers (a 2-pool DNA panel and 1-pool RNA panel) and Genexus Strips 1 & 2-AS library reagents
    • Enough to sequence up to 32 samples on the Genexus Integrated Sequencer
  • DNA panel is comprised of 45 key genes, while the RNA panel includes a broad fusion panel of 30 driver genes, covering the most relevant targets associated with major myeloid disorders
  • Comprehensive coverage of key DNA mutations and >700 fusion transcripts associated with myeloid disorders
  • Sequencing of up to eight samples (DNA & RNA) per lane on a GX5 Chip in a single run
  • Less than 15 minutes hands-on time
  • Detection of somatic variants down to 5% allele frequency
  • Depending on the workflow, results can be obtained in as little as a single day
  • For research use only, not for use in diagnostic procedures

Spécifications

Compatibilité de puce GX5
Contenu et stockage Store at -30°C to -10°C.
Type d’échantillon Whole Blood, Bone Marrow
Type de séquençage Next Generation Sequencing
À utiliser avec (équipement) Genexus Integrated Sequencer
À utiliser avec (application) Oncology Research
Bibliothèques Amplicon
Type d’acide nucléique DNA and RNA
Nb d’échantillons Up to 32 Samples
Gamme de produit Oncomine
Type de produit Assay
Quantité 32 samples
Débit Up to 8 DNA+RNA Samples per GX5 lane
Étape du flux de travail Panel and Library
Afficher plus de résultats Afficher moins de résultats