Learn More
Diba Omnifit™ T-Series Solvent Waste Caps with Vent from Cole-Parmer™
Catalog No. 14500717
Change view
Click to view available options
Includes:
4 Luer Ports
Catalog No. Includes
14500717 4 Luer Ports
1 options
Catalog No. 14500717 Supplier Diba Supplier No. 00983T 4W
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Connect a vapor trap using the Diba™ Omnifit™ T-Series Solvent Waste Cap's vent port to prevent solvent vapors from escaping.
  • Use without any fittings or use male Luer fittings to connect various tubing sizes
  • Anti-twist design
  • Autoclavable
  • 4W - White inner body with blue outer cap
  • 8W - White inner body and outer cap
Specifications
Includes 4 Luer Ports
Material PTFE inner body/PP outer cap
Thread Size 83B
Product Type Cap