Learn More
Thermo Scientific™ Olympus™ 1.25X Objective, PlanApo N, 0.04 NA
Catalog No. N010125
Encompass
Change view
Click to view available options
Quantity:
1 each
Catalog No. Quantity
N010125 1 each
1 options
Catalog No. N010125 Supplier Thermo Scientific™ Supplier No. N010125
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Expand the functionality of your Thermo Scientific™ high-content instrument
Add a wide range of magnifications using Carl Zeiss™ and Olympus™ Objective Lenses for Thermo Scientific™ ArrayScan™ HCA Readers and the Thermo Scientific™ CellInsight™ NXT High Content Screening (HCS) Platform.
Specifications
Type Objective Lens
Product Line Olympus
Description Olympus™ 1.25X Objective, PlanApo N (0.04 NA)
For Use With (Equipment) ArrayScan HCA Readers
Lens Type Dry
Magnification 1.25x (0.04NA)
Quantity 1 each
For Research Use Only. Not for use in diagnostic procedures.