missing translation for 'onlineSavingsMsg'
Learn More

Olmesartan Medoxomil Related Compound A Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Numéro de catalogue. 111015681
Modifier l'affichage
Click to view available options
Quantity:
40 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
111015681 40 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 111015681 Fournisseur Merck Emd Millipore Code fournisseur PHR201640MG
Il en reste null
Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Olmesartan Medoxomil Related Compound A is found as an impurity of olmesartan medoxomil. Olmesartan medoxomil is a specific angiotensin II type 1 antagonist, which is widely used as an antihypertensive agent for the treatment of hypertension.

Spécifications

CAS 849206-43-5
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C24H24N6O2
Synonym 3,6-Dihydro-6,6-dimethyl-2-propyl-3-[[2′-(2 H-tetrazol-5-yl)[1,1′-biphenyl]-4-yl]methyl]-4 H-furo[3,4-d]imidazol-4-one; 1-{[2′-(1 H-Tetrazol-5-yl)biphenyl-4-yl]methyl}-4,4-dimethyl-2-propyl-1 H-furo[3,4-d]imidazol-6(4 H)-one
Type Certified Reference Material
Quantity 40 mg
Formula Weight 428.49
Grade Certified Reference Material