missing translation for 'onlineSavingsMsg'
Learn More

Olive oil, pure, refined

Catalog No. AC416535000
Change view
Click to view available options
Quantity:
25 mL
500 mL
2.5 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC416535000 500 mL Glass bottle
AC416530250 25 mL Glass bottle
AC416530025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC416535000 Supplier Thermo Scientific Chemicals Supplier No. 416535000

CAS: 8001-25-0

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 8001-25-0
MDL Number MFCD00131764

Specifications

Acidity 0.3 max. (mg KOH/g)
Absorbance 1.20 max. (1g/100ml cyclohexane) measured at the maximum between 264nm and 276nm
Color Colorless or Green-Yellow
Density 0.9135g/mL
Flash Point 225°C
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4670 to 1.4710
Quantity 500 mL
Specific Gravity 0.9135
Merck Index 14, 6836
Solubility Information Solubility in water: .. Other solubilities: practically insoluble in alcohol, miscible with light petroleum (50-60°C)
Grade Pure
Concentration or Composition (by Analyte or Components) of fatty acids: 0.1% max. (< C16), of the sterol fraction: Sterols: min. 93.0%, Cholesterol: max. 0.5%, Campesterol: max. 4.0%, delta 7-Stigmasterol: max. 0.5%
Physical Form Viscous Liquid
Chemical Name or Material Olive oil
Show More Show Less
EINECSNumber 232-277-0
RTECSNumber RK4300000
TSCA TSCA

RUO – Research Use Only