missing translation for 'onlineSavingsMsg'
Learn More

Olive oil, MP Biomedicals™

Catalog No. ICN10250290
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10250290 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10250290 Supplier MP Biomedicals Inc Supplier No. 0210250290

Chemical Identifiers

CAS 8001-25-0

Specifications

Color Green to Yellow
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Olive oil