missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Oligomer A11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. AHB0052
Encompass
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AHB0052 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AHB0052 Supplier Invitrogen™ Supplier No. AHB0052
Only null left

Rabbit Polyclonal Antibody

This antibody recognizes amino acid sequence-independent oligomers of proteins or peptides. A11 does not recognize monomers or mature fibers of proteins or peptides. A11 reacts with soluble AB42 oligomers and does not react with soluble low molecular weight AB40 or AB40 fibrils. A11 recognizes oligomeric species of several other amyloidogenic polypeptides including AB42, human insulin, prion, polyglutamine, lysozyme, alpha-synuclein and yeast prion Sup35. Oligomeric AB42 is recommended as a positive control, monomeric and fibrillar AB42 as negative controls.

Many degenerative diseases are known to be related to the accumulation of misfolded proteins called, amyloid fibrils. Soluble amyloid oligomers are considered as the principal pathogenic species which play an important role in the formation of amyloid fibrils; therefore, oligomers are involved in the pathogenesis of the many neurodegenerative diseases. This anti-oligomer antibody provides a facile means of assessing the significance of oligomers in disease pathogenesis.

Specifications

Antigen Oligomer A11
Applications Dot Blot, ELISA, Immunohistochemistry, Neutralization, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene INS
Gene Accession No. P01308, P01322, P01325
Gene Alias Antioligomer A11; IDDM; IDDM1; IDDM2; ILPR; INS; Ins1; Ins-1; Ins2-rs1; Insulin; insulin 1; Insulin A chain; Insulin B chain; insulin I; insulin-1; Insulin-1 A chain; Insulin-1 B chain; IRDN; MODY10; preproinsulin; proinsulin
Gene Symbols INS, Ins1
Host Species Rabbit
Immunogen Synthetic molecular mimic of soluble oligomers.
Purification Method Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16333, 24505, 3630
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less