missing translation for 'onlineSavingsMsg'
Learn More

Oleic Acid 99.0+%, TCI America™

Catalog No. O01805ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O01805ML 5 mL
O018025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O01805ML Supplier TCI America Supplier No. O01805ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 112-80-1
Molecular Formula C18H34O2
Molecular Weight (g/mol) 282.47
MDL Number MFCD00064242
InChI Key ZQPPMHVWECSIRJ-MDZDMXLPSA-N
Synonym oleic acid, cis-9-octadecenoic acid, cis-oleic acid, elaidoic acid, oleate, glycon wo, wecoline oo, pamolyn 100, glycon ro, z-octadec-9-enoic acid
PubChem CID 445639
ChEBI CHEBI:16196
IUPAC Name (9E)-octadec-9-enoic acid
SMILES CCCCCCCC\C=C\CCCCCCCC(O)=O

Specifications

Color Yellow
Boiling Point 285°C
Quantity 5 mL
Formula Weight 282.47
Percent Purity ≥99.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Oleic Acid
EINECSNumber (2)-0609&(2)-0975
RTECSNumber RG2275000
TSCA Yes
Recommended Storage Refrigerator