missing translation for 'onlineSavingsMsg'
Learn More

Oleic Acid 85.0+%, TCI America™

Catalog No. O0011500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O0011500ML 500 mL
O001125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O0011500ML Supplier TCI America Supplier No. O0011500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 112-80-1
Molecular Formula C18H34O2
Molecular Weight (g/mol) 282.47
MDL Number MFCD00064242
InChI Key ZQPPMHVWECSIRJ-MDZDMXLPSA-N
Synonym oleic acid, cis-9-octadecenoic acid, cis-oleic acid, elaidoic acid, oleate, glycon wo, wecoline oo, pamolyn 100, glycon ro, z-octadec-9-enoic acid
PubChem CID 445639
ChEBI CHEBI:16196
IUPAC Name (9E)-octadec-9-enoic acid
SMILES CCCCCCCC\C=C\CCCCCCCC(O)=O

Specifications

Color Yellow
Boiling Point 285°C
Quantity 500 mL
Formula Weight 282.47
Percent Purity ≥85.0% (GC)
Physical Form Liquid
Chemical Name or Material Oleic Acid
EINECSNumber (2)-0609&(2)-0975
RTECSNumber RG2275000
TSCA Yes
Recommended Storage Refrigerator