missing translation for 'onlineSavingsMsg'
Learn More

Oleic Acid, Purified, (cis-9-Octadecenoic Acid), Reagents

Catalog No. 01337583
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337583 100 mL
01337582 1 L
01337581 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01337583 Supplier Labchem Inc Supplier No. C2110000100C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Oleic Acid, Purified, (cis-9-Octadecenoic Acid), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Oleic Acid, Purified, (cis-9-Octadecenoic Acid)
CAS 112-80-1
Boiling Range 286°C
Melting Point 14°C
Color Colorless to Slight Yellow
Boiling Point 286°C
Melting Range 14°C
Packaging Glass Bottle
Molecular Formula C18H34O2
Quantity 100 mL
Specific Gravity 0.895
Synonym cis-9-Octadecenoic Acid
Molecular Weight (g/mol) 282.2559
Grade Reagent
Physical Form Liquid
Show More Show Less