missing translation for 'onlineSavingsMsg'
Learn More

Oleic Acid Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012508
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012508 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012508 Supplier Merck Emd Millipore Supplier No. PHR15861G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Specifications

CAS 112-80-1
Vapor Pressure 1 mmHg (176°C)
Density 0.89 g/mL (at 25°C)
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula CH3(CH2)7 CH=CH(CH2)7 COOH
MDL Number MFCD00064242
Melting Point 13°C to 14°C
Boiling Point 194°C to 195°C (1.2 mmHg)
Synonym Oleic acid; cis-9-Octadecenoic acid; Elainic acid
Type Certified Reference Material
Quantity 1 g
Formula Weight 282.46
Grade Certified Reference Material
Show More Show Less