missing translation for 'onlineSavingsMsg'
Learn More

Oleic Acid, Food Grade, Spectrum™ Chemical

Catalog No. 18604881
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604881 20 L Amber Glass Bottle
18604878 1 L Amber Glass Bottle
18604879 4 L Amber Glass Bottle
18604880 200 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18604881 Supplier Spectrum Chemical Mfg Cor Supplier No. O150020LT
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

O1500, 112-80-1, C18H34O2

Spectrum™ Chemical Oleic Acid, Food Grade

Chemical Identifiers

CAS 112-80-1
Molecular Formula C18H34O2
Molecular Weight (g/mol) 282.47
MDL Number MFCD00064242
InChI Key ZQPPMHVWECSIRJ-MDZDMXLPSA-N
IUPAC Name (9E)-octadec-9-enoic acid
SMILES CCCCCCCC\C=C\CCCCCCCC(O)=O

Specifications

CAS Max % 1
Boiling Point 286.1°C
Packaging Amber Glass Bottle
Quantity 20 L
Saponification Value 196-204
Grade Food Grade
Physical Form Liquid