missing translation for 'onlineSavingsMsg'
Learn More

Oleanolic acid, ≥97.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014380
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014380 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014380 Supplier Merck Emd Millipore Supplier No. 4251510MG
Only null left
Food and Beverages

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 508-02-1
Physical Form Neat
Linear Formula C30H48O3
MDL Number MFCD00064914
Melting Point >300°C
Type Analytical Standard
Quantity 10 mg
Formula Weight 456.70
Percent Purity ≥97.0% (HPLC)
Grade Analytical Standard
Show More Show Less