missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Olaparib

Catalog No. AC466290010
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC466290010 1 g
AC466292500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC466290010 Supplier Thermo Scientific Chemicals Supplier No. 466290010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 763113-22-0 | C24H23FN4O3, C24H23FN4O3 | 434.47 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 763113-22-0
Molecular Formula C24H23FN4O3

Specifications

Quantity 1 g
Melting Point 198.0°C
Formula Weight 434.47
InChI Key FDLYAMZZIXQODN-UHFFFAOYSA-N
SMILES FC1=C(C=C(CC2=NNC(=O)C3=CC=CC=C23)C=C1)C(=O)N1CCN(CC1)C(=O)C1CC1
IUPAC Name 4-{[3-(4-cyclopropanecarbonylpiperazine-1-carbonyl)-4-fluorophenyl]methyl}-1,2-dihydrophthalazin-1-one
Molecular Weight (g/mol) 434.47
Chemical Name or Material Olaparib
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Toxic if swallowed.

May damage the unborn child.

Causes damage to organs through prolonged or repeated exposure if inhaled.

Precautionary Statement GHS P Statement Wash face, hands and any exposed skin thoroughly after handling.

IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Obtain special instructions before use.

Wear protective gloves/protective clothing/eye protection/face protection.

IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only