CAS RN 763113-22-0
IUPAC Name:
4-{[3-(4-cyclopropanecarbonylpiperazine-1-carbonyl)-4-fluorophenyl]methyl}-1,2-dihydrophthalazin-1-one
Synonyms:
olaparib
Molecular Weight (g/mol):
434.47
Molecular Formula:
C24H23FN4O3
InChi Key:
FDLYAMZZIXQODN-UHFFFAOYSA-N
SMILES:
FC1=C(C=C(CC2=NNC(=O)C3=CC=CC=C23)C=C1)C(=O)N1CCN(CC1)C(=O)C1CC1
1
–
2
of
2
results
1
Promotions Available
Thermo Scientific Chemicals Olaparib
Thermo Scientific Chemicals Olaparib
CAS: 763113-22-0 Molecular Formula: C24H23FN4O3
2
R&D Systems™ Olaparib
A Tocris product, part of R&D Systems. Potent PARP inhibitor