missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Olanzapine

Numéro de catalogue. 434910
Modifier l'affichage
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
434910 10 mg
434950 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. 434910 Fournisseur Tocris Bioscience™ Code fournisseur 4349/10
Il en reste null

5-HT2A/D2 antagonist; atypical antipsychotic

Antagonist of 5-HT2A and dopamine D2 receptors (Ki values are 4 and 11 nM respectively). Also displays affinity for a range of other receptors including D1 and D4, 5-HT2C, α1, H1 and M1-4 receptors (Ki values range between 1.9 and 31 nM) . Atypical antipsychotic. Displays anticholinergic properties. Also a highly potent activator of hM4Di DREADDs (EC50 values are 5 nM in vitro and 7 nM in vivo).

Identifiants chimiques

CAS 132539-06-1
Molecular Formula C17H20N4S

Spécifications

Quantity 10 mg
Formula Weight 312.43
Purity >99%
Molecular Weight (g/mol) 312.44
Chemical Name or Material Olanzapine
Recommended Storage Store at +4°C