missing translation for 'onlineSavingsMsg'
Learn More

Octylmagnesium chloride, 1.4M solution in THF, AcroSeal™, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 428038000

 View more versions of this product

Catalog No. AC428038000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Octylmagnesium chloride
38841-98-4 , 109-99-9
29.79,77.66
C8H17ClMg
MFCD00000476
octylmagnesium chloride, magnesium, chlorooctyl, n-octylmagnesiumchloride, octyl magnesium chloride, n-c8h17mgcl, n-octylmagnesium chloride, octylmagnesium chloride solution, dpbwwqdkvqlwfk-uhfffaoysa-m, 1-octylmagnesium chloride, 1m in methf, octylmagnesium chloride solution, 2.0 m in thf
CCCCCCC[CH2-].[Mg+2].[Cl-]
172.98
172.98
0.929
1.4M solution in THF
22.34,70.21
800mL
CH3(CH2)6CH2MgCl
−10°C
HQDAZWQQKSJCTM-UHFFFAOYSA-M
magnesium;octane;chloride
4147387
AcroSeal™ Glass bottle
0.9290g/mL

Chemical Identifiers

38841-98-4
172.98
HQDAZWQQKSJCTM-UHFFFAOYSA-M
4147387
CCCCCCC[CH2-].[Mg+2].[Cl-]
C8H17ClMg
MFCD00000476
octylmagnesium chloride, magnesium, chlorooctyl, n-octylmagnesiumchloride, octyl magnesium chloride, n-c8h17mgcl, n-octylmagnesium chloride, octylmagnesium chloride solution, dpbwwqdkvqlwfk-uhfffaoysa-m, 1-octylmagnesium chloride, 1m in methf, octylmagnesium chloride solution, 2.0 m in thf
magnesium;octane;chloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
May form explosive peroxides.
Reacts violently with water.
Suspected of causing cancer.<br/

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

GHS Signal Word: Danger

EINECSNumber : 254-147-2

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only