accessibility menu, dialog, popup

UserName

H3CMgCl

CAS RN 38841-98-4

IUPAC Name: chloro(octyl)magnesium
Synonyms: chloro(octyl)magnesium
Molecular Weight (g/mol): 172.98
Molecular Formula: C8H17ClMg
InChi Key: DPBWWQDKVQLWFK-UHFFFAOYSA-M
SMILES: CCCCCCCC[Mg]Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results