missing translation for 'onlineSavingsMsg'
Learn More

Octylmagnesium chloride, 1.4M solution in THF, AcroSeal™

Catalog No. p-4089938
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC428031000 100 mL AcroSeal™ Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC428031000 Supplier Thermo Scientific Chemicals Supplier No. 428031000
Only null left

CAS: 38841-98-4 | C8H17ClMg | 172.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 38841-98-4 , 109-99-9
Molecular Formula C8H17ClMg
Molecular Weight (g/mol) 172.98
MDL Number MFCD00000476
InChI Key DPBWWQDKVQLWFK-UHFFFAOYSA-M
Synonym octylmagnesium chloride, magnesium, chlorooctyl, n-octylmagnesiumchloride, octyl magnesium chloride, n-c8h17mgcl, n-octylmagnesium chloride, octylmagnesium chloride solution, dpbwwqdkvqlwfk-uhfffaoysa-m, 1-octylmagnesium chloride, 1m in methf, octylmagnesium chloride solution, 2.0 m in thf
PubChem CID 4147387
SMILES CCCCCCCC[Mg]Cl

Specifications

CAS Min % 22.34, 70.21
CAS Max % 29.79, 77.66
Quantity 100 mL
Linear Formula CH3(CH2)6CH2MgCl
Flash Point −10°C
IUPAC Name magnesium;octane;chloride
Formula Weight 172.98
Packaging AcroSeal™ Glass bottle
Specific Gravity 0.929
Density 0.9290g/mL
Chemical Name or Material Octylmagnesium chloride, 1.4M solution in THF
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
May form explosive peroxides.
Reacts violently with water.
Suspected of causing cancer.<br/
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p
EINECSNumber 254-147-2

RUO – Research Use Only