missing translation for 'onlineSavingsMsg'
Learn More

Octyldodecanol Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014498
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014498 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014498 Supplier Merck Emd Millipore Supplier No. PHR11551G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Specifications

CAS 5333-42-6
Density 0.838 g/mL (at 25°C)
Flash Point 188°C - open cup
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula CH3(CH2)9 CH[(CH2)7 CH3 ]CH2 OH
MDL Number MFCD01310428
Melting Point -1°C to +1°C
Boiling Point 234°C to 238°C (33 mmHg)
Synonym 2-Octyl-1-dodecanol
Type Certified Reference Material
Quantity 1 g
Formula Weight 298.55
Grade Certified Reference Material
Show More Show Less