Octet SPR Sample Rack Seal Pack of 5. Octet SPR Plate Seals are self-adhesive transparent microfoils compatible with polystrene and polypropylene plates and are designed to prevent any sample or reagent evaporation during long SPR assay run times.
Numéro de catalogue.
502891627
Fournisseur
Danaher CorpCode fournisseur
190103
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Octet SPR Sample Rack Seal Pack of 5. Octet SPR Plate Seals are self-adhesive transparent microfoils compatible with polystrene and polypropylene plates and are designed to prevent any sample or reagent evaporation during long SPR assay run times.