Octet SPR Plate Seal Pack of 5 self-adhesive transparent microfoils compatible with polystrene and polypropylene plates and are designed to prevent any sample or reagent evaporation during long SPR assay run times.
Numéro de catalogue.
502891688
Fournisseur
Danaher CorpCode fournisseur
190102
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Octet SPR Plate Seal Pack of 5 self-adhesive transparent microfoils compatible with polystrene and polypropylene plates and are designed to prevent any sample or reagent evaporation during long SPR assay run times.