accessibility menu, dialog, popup

UserName

H3COHO

CAS RN 124-07-2

IUPAC Name: octanoic acid
Synonyms: caprylic acid zinc salt caprylic acid lanthanum(+3) salt caprylic acid caprylic acid, 14c-labeled lead(+2) salt caprylic acid cobalt salt caprylic acid kortacid 0899 lithium salt caprylic acid aliphatic carboxylic acid, c8:0 acide octanoique
Molecular Weight (g/mol): 144.21
Molecular Formula: C8H16O2
InChi Key: WWZKQHOCKIZLMA-UHFFFAOYSA-N
SMILES: CCCCCCCC(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (5)
  • (7)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
Boiling Point 
  • (4)
  • (2)
  • (3)
  • (5)
Percent Purity 
  • (4)
  • (2)
  • (5)
  • (3)
Physical Form 
  • (11)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (12)
  • (5)
  • (1)
  • (5)
  • (2)
  • (7)
  • (3)
  • (5)
  • (7)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (4)
  • (2)
  • (3)
  • (5)
  • (4)
  • (2)
  • (5)
  • (3)
  • (11)
  • (5)
  • (1)