missing translation for 'onlineSavingsMsg'
Learn More

Octanoic Acid, 97%, Spectrum™ Chemical

Catalog No. 18612072
Change view
Click to view available options
Quantity:
100 mL
1 L
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612072 100 mL Amber Glass Bottle
18612071 1 L Amber Glass Bottle
18612075 4 L Amber Glass Bottle
18612073 20 L Amber Glass Bottle
18612074 200 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18612072 Supplier Spectrum Chemical Mfg Cor Supplier No. O1003100ML
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 124-07-2
Molecular Weight (g/mol) 144.21 g/mol

Specifications

CAS Max % 1
Color Colorless to Light Yellow
Boiling Point 239°C
Packaging Amber Glass Bottle
Quantity 100 mL
Percent Purity 0.97
Grade Reagent
Physical Form Liquid
Chemical Name or Material Octanoic Acid

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.