missing translation for 'onlineSavingsMsg'
Learn More

Octane-1-sulfonic acid sodium salt, LiChropur™ EMD-Millipore

Catalog No. M1183070025
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1183070025 25 g Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1183070025 Supplier Technidata Supplier No. 1.18307.0025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ion pair reagents for analytical HPLC LiChropur

Chemical Identifiers

CAS 5324-84-5
Molecular Formula C8H17NaO3S
Molecular Weight (g/mol) 216.27
MDL Number MFCD00007544
InChI Key HRQDCDQDOPSGBR-UHFFFAOYSA-M
Synonym sodium 1-octanesulfonate, sodium octane-1-sulfonate, 1-octanesulfonic acid sodium salt, sodium octanesulfonate, 1-octanesulfonic acid, sodium salt, 1-octanesulfonate, sodium, sodium octylsulfonate, unii-j46ez5jyb3, octyl sodium sulfonate, ipc-alks-8
PubChem CID 23669624
IUPAC Name sodium octane-1-sulfonate
SMILES [Na+].CCCCCCCCS([O-])(=O)=O

Specifications

Density 345kg/m3
Packaging Glass
Quantity 25 g
Formula Weight 216.27
Percent Purity ≥99%
Grade Not Available
Physical Form Solid
Chemical Name or Material Octane-1-sulfonic acid sodium salt
Recommended Storage Store at 5° to 30°C