missing translation for 'onlineSavingsMsg'
Learn More

Octanal, 98%

Catalog No. AAA1090130
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1090130 250 g
AAA1090118 50 g
AAA109010B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1090130 Supplier Thermo Scientific Chemicals Supplier No. A1090130
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 124-13-0 | C8H16O | 128.22 g/mol

Octanal is an aromatic aldehyde often found in citrus oils. It is used commercially as a component in perfumes and in flavor production for the food industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Octanal is an aromatic aldehyde often found in citrus oils. It is used commercially as a component in perfumes and in flavor production for the food industry.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Handle and store under inert gas. Air and moisture sensitive. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 124-13-0
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.22
MDL Number MFCD00007029
InChI Key NUJGJRNETVAIRJ-UHFFFAOYSA-N
Synonym caprylic aldehyde, caprylaldehyde, n-octanal, 1-octanal, n-octyl aldehyde, n-octaldehyde, n-caprylaldehyde, octanaldehyde, aldehyde c-8, n-octylal
PubChem CID 454
ChEBI CHEBI:17935
IUPAC Name octanal
SMILES CCCCCCCC=O

Specifications

Melting Point 12°C to 15°C
Density 0.821
Boiling Point 169°C to 171°C
Flash Point 56°C (133°F)
Refractive Index 1.4195
Linear Formula CH3(CH2)6CHO
Quantity 250 g
UN Number UN1191
Sensitivity Air Sensitive
Merck Index 14,1766
Solubility Information Slightly soluble in water.
Formula Weight 128.22
Percent Purity 98%
Chemical Name or Material Octanal
Show More Show Less
Hazard Category H226-H315-H319
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P332+P313-P363-P370+P378q-P374-P380-P390-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: OCTYL ALDEHYDES
EINECSNumber 204-683-8
RTECSNumber RG7780000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only