missing translation for 'onlineSavingsMsg'
Learn More

Octafluorotoluene 98.0+%, TCI America™

Catalog No. P08565G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P08565G 5 g
P085625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P08565G Supplier TCI America Supplier No. P08565G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 434-64-0
Molecular Formula C7F8
Molecular Weight (g/mol) 236.06
MDL Number MFCD00000375
InChI Key USPWUOFNOTUBAD-UHFFFAOYSA-N
Synonym octafluorotoluene, perfluorotoluene, 1,2,3,4,5-pentafluoro-6-trifluoromethyl benzene, toluene, octafluoro, benzene, pentafluoro trifluoromethyl, trifluoromethyl pentafluorobenzene, pentafluorobenzotrifluoride, 1,2,3,4,5-pentafluoro-6-trifluoromethyl-benzene, benzene, 1,2,3,4,5-pentafluoro-6-trifluoromethyl, acmc-209juh
PubChem CID 9906
IUPAC Name 1,2,3,4,5-pentafluoro-6-(trifluoromethyl)benzene
SMILES FC1=C(F)C(F)=C(C(F)=C1F)C(F)(F)F

Specifications

Color Yellow
Boiling Point 104°C
Quantity 5 g
UN Number 1993
Formula Weight 236.06
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Octafluorotoluene
RTECSNumber XT3530000
TSCA Yes