missing translation for 'onlineSavingsMsg'
Learn More

Octafluoro-1,4-diiodobutane (stabilized with Copper chip) 98.0+%, TCI America™

Catalog No. D23295G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D23295G 5 g
D232925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D23295G Supplier TCI America Supplier No. D23295G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 375-50-8
Molecular Formula C4F8I2
Molecular Weight (g/mol) 453.84
MDL Number MFCD00042263
InChI Key JILAKKYYZPDQBE-UHFFFAOYSA-N
Synonym 1,4-diiodooctafluorobutane, octafluoro-1,4-diiodobutane, 1,4-diiodoperfluorobutane, perfluoro-1,4-diiodobutane, 1,4-diiodo octafluorobutane, butane, 1,1,2,2,3,3,4,4-octafluoro-1,4-diiodo, acmc-1aigi, i cf2 4i, 1,1,2,2,3,3,4,4-octafluoro-1,4-diiodo-butane
PubChem CID 78993
IUPAC Name 1,1,2,2,3,3,4,4-octafluoro-1,4-diiodobutane
SMILES FC(F)(I)C(F)(F)C(F)(F)C(F)(F)I

Specifications

Boiling Point 63°C
Quantity 5 g
Formula Weight 453.84
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Octafluoro-1,4-diiodobutane (stabilized with Copper chip)
EINECSNumber (2)-4014
TSCA No