missing translation for 'onlineSavingsMsg'
Learn More

Octaethylene Glycol Monododecyl Ether 97.0+%, TCI America™

Catalog No. O01395G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O01395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O01395G Supplier TCI America Supplier No. O01395G

Chemical Identifiers

CAS 3055-98-9
Molecular Formula C28H58O9
Molecular Weight (g/mol) 538.763
MDL Number MFCD00043234
InChI Key YYELLDKEOUKVIQ-UHFFFAOYSA-N
Synonym octaethylene glycol monododecyl ether, dodecyloctaethyleneglycol monoether, octaethyleneglycol monododecyl ether, 3,6,9,12,15,18,21,24-octaoxahexatriacontan-1-ol, octaethyleneglycol-dodecylmonoether, o-dodecanyl octaethylene glycol, n-dodecyl octaethylene glycol monoether, octaethylene glycol mono-n-dodecyl ether, ce1, dodecyloctaglycol
PubChem CID 123921
ChEBI CHEBI:41527
IUPAC Name 2-[2-[2-[2-[2-[2-[2-(2-dodecoxyethoxy)ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethanol
SMILES CCCCCCCCCCCCOCCOCCOCCOCCOCCOCCOCCOCCO

Specifications

Color White
Quantity 5 g
Formula Weight 538.76
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Octaethylene Glycol Monododecyl Ether
EINECSNumber (7)-0097
TSCA No