missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Octadecylamine Hydrochloride 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 162°C |
| Color | White-Yellow |
| Quantity | 25 g |
| Formula Weight | 305.98 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Octadecylamine Hydrochloride |
Safety and Handling
| RTECSNumber | RG4375000 |
| TSCA | Yes |