missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ OAZ2 Polyclonal Antibody
GREENER_CHOICE

Numéro de catalogue. PIPA5103991
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
PIPA5103991 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. PIPA5103991 Fournisseur Invitrogen™ Code fournisseur PA5103991
Il en reste null

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total OAZ2.

Ornithine decarboxylase catalyzes the conversion of ornithine to putrescine in the first and apparently rate-limiting step in polyamine biosynthesis. The ornithine decarboxylase antizymes play a role in the regulation of polyamine synthesis by binding to and inhibiting ornithine decarboxylase. Antizyme expression is auto-regulated by polyamine-enhanced translational frameshifting. The antizyme encoded by this gene inhibits ornithine decarboxylase but does not accelerate its degradation.

Spécifications

Antigen OAZ2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene OAZ2
Gene Accession No. O08608, O95190
Gene Alias antizyme 2; AZ2; AZ-2; OAZ2; Oaz2-ps; ODC-Az 2; ornithine decarboxylase antizyme 2; RGD1562933; seizure-related protein 15; Sez15
Gene Symbols OAZ2
Host Species Rabbit
Immunogen A synthesized peptide derived from human OAZ2(Accession O95190), corresponding to amino acid residues L119-P169.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18247, 4947, 501454
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Afficher plus de résultats Afficher moins de résultats