Learn More
Waters Limited Oasis MCX 20 cc Vac RC Cartridge, 60 mg Sorbent per Cartridge, 30 m, 50/pk
Numéro de catalogue. 50780154
Encompass
These Vac RC cartridge contains the Oasis MCX sorbent, which is a novel, mixed-mode polymeric sorbent that has been optimized to achieve higher selectivity and sensitivity for extracting basic compounds with cation-exchange groups. Since the Oasis MCX sorbent is water wettable, it maintains its capability for higher retention and excellent recoveries even if the sorbent runs dry, which means there is no need to take extraordinary steps to keep the sorbent beds from drying out during the critical steps prior to sample loading. These cartridges feature an integral 20 cc funnel-shaped reservoir for convenient processing of greater volumes or samples or solvents. They can be used with robotic sample preparation equipment or with vacuum manifolds.
Numéro de catalogue. 50780154 Fournisseur Waters Limited Code fournisseur 186000261
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
These Vac RC cartridge contains the Oasis MCX sorbent, which is a novel, mixed-mode polymeric sorbent that has been optimized to achieve higher selectivity and sensitivity for extracting basic compounds with cation-exchange groups. Since the Oasis MCX sorbent is water wettable, it maintains its capability for higher retention and excellent recoveries even if the sorbent runs dry, which means there is no need to take extraordinary steps to keep the sorbent beds from drying out during the critical steps prior to sample loading. These cartridges feature an integral 20 cc funnel-shaped reservoir for convenient processing of greater volumes or samples or solvents. They can be used with robotic sample preparation equipment or with vacuum manifolds.