missing translation for 'onlineSavingsMsg'
Learn More
Learn More
o-Xylene, Reagent Grade, ≥98.0%, Honeywell
Reagent Grade,≥98.0%
$1471.83 - $1471.83
Chemical Identifiers
| CAS | 95-47-6 |
|---|---|
| Molecular Formula | C8H10 |
| Molecular Weight (g/mol) | 106.17 |
| MDL Number | MFCD00008519 |
| InChI Key | CTQNGGLPUBDAKN-UHFFFAOYSA-N |
| Synonym | o-xylene, 1,2-dimethylbenzene, ortho-xylene, o-xylol, o-methyltoluene, o-dimethylbenzene, 2-xylene, 3,4-xylene, benzene, 1,2-dimethyl, o-xylenes |
| PubChem CID | 7237 |
| ChEBI | CHEBI:28063 |
| IUPAC Name | 1,2-xylene |
| SMILES | CC1=CC=CC=C1C |
Description
- ≥95% for general laboratory use.
Chemical Identifiers
| 95-47-6 | |
| 106.17 | |
| CTQNGGLPUBDAKN-UHFFFAOYSA-N | |
| 7237 | |
| 1,2-xylene |
| C8H10 | |
| MFCD00008519 | |
| o-xylene, 1,2-dimethylbenzene, ortho-xylene, o-xylol, o-methyltoluene, o-dimethylbenzene, 2-xylene, 3,4-xylene, benzene, 1,2-dimethyl, o-xylenes | |
| CHEBI:28063 | |
| CC1=CC=CC=C1C |
Specifications
| 95-47-6 | |
| 18 L | |
| 31°C (87.8°F) | |
| C8H10 | |
| MFCD00008519 | |
| UN1307 | |
| o-xylene, 1,2-dimethylbenzene, ortho-xylene, o-xylol, o-methyltoluene, o-dimethylbenzene, 2-xylene, 3,4-xylene, benzene, 1,2-dimethyl, o-xylenes | |
| CC1=CC=CC=C1C | |
| 106.17 | |
| CHEBI:28063 | |
| ≥98% |
| 0.879g/mL at 20°C | |
| 0.07 | |
| Stainless Steel Can | |
| C6H4(CH3)2 | |
| 16 mmHg (37.7°C), 0.1 atm (21.1°C), 7 mmHg (20°C) | |
| 1815558 | |
| CTQNGGLPUBDAKN-UHFFFAOYSA-N | |
| 1,2-xylene | |
| 7237 | |
| 106.17g/mol | |
| o-Xylene |
Safety and Handling
P210-P261-P273-P301 + P310-P302 + P352 + P312-P331
RTECSNumber : ZE2450000