missing translation for 'onlineSavingsMsg'
Learn More
Learn More
o-Vanillin 99.0+%, TCI America™
Specifications
Specifications
| Melting Point | 44°C |
| Boiling Point | 268°C |
| Quantity | 100 g |
| Formula Weight | 152.15 |
| Percent Purity | ≥99.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | o-Vanillin |
Safety and Handling
| EINECSNumber | (3)-1200 |
| RTECSNumber | CU6530000 |
| TSCA | Yes |