missing translation for 'onlineSavingsMsg'
Learn More

o-Tolylmagnesium bromide, 2M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals

Catalog No. AC427471000
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC427471000 100 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC427471000 Supplier Thermo Scientific Chemicals Supplier No. 427471000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 932-31-0 | Molecular Formula: C7H7BrMg | Molecular Weight: 195.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 932-31-0 , 60-29-7
Molecular Formula C7H7BrMg
Molecular Weight (g/mol) 195.34
MDL Number MFCD00010350
InChI Key DVXDIGKNJYSMFM-UHFFFAOYSA-M
Synonym o-tolylmagnesium bromide, bromo-o-tolylmagnesium, bromo 2-methylphenyl magnesium, 2-tolylmagnesium bromide, o-methylphenylmagnesium bromide, 2-methylphenylmagnesium bromide, magnesium,bromo 2-methylphenyl, o-tolyl magnesium bromide, tolylmagnesium bromide, o-tolylmagnesiumbromid, grignard reagent
PubChem CID 101919
IUPAC Name magnesium;methylbenzene;bromide
SMILES CC1=CC=CC=C1[Mg]Br

Specifications

Color Brown to Gray
Density 1.0130g/mL
Flash Point −40°C
Packaging AcroSeal™ Glass Bottle
Linear Formula CH3C6H4MgBr
Quantity 100 mL
Specific Gravity 1.013
Formula Weight 195.34
Physical Form Solution
Chemical Name or Material o-Tolylmagnesium bromide, 2M solution in diethyl ether
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
Harmful if swallowed.
Extremely flammable liquid and vapor.
May form explosive peroxides.
Reacts violently with water.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 213-250-2

RUO – Research Use Only