missing translation for 'onlineSavingsMsg'
Learn More

o-Tolyl Trifluoromethanesulfonate 98.0+%, TCI America™

Catalog No. T33575G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T33575G 5 g
T335725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T33575G Supplier TCI America Supplier No. T33575G
Only null left

Chemical Identifiers

CAS 66107-34-4
Molecular Formula C8H7F3O3S
Molecular Weight (g/mol) 240.20
MDL Number MFCD29089375
InChI Key RBOGJRXPKONDGC-UHFFFAOYSA-N
Synonym Trifluoromethanesulfonic Acid o-Tolyl Ester, o-Tolyl Triflate
PubChem CID 572025
IUPAC Name 2-methylphenyl trifluoromethanesulfonate
SMILES CC1=CC=CC=C1OS(=O)(=O)C(F)(F)F

Specifications

Color Colorless
Quantity 5 g
Formula Weight 240.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material o-Tolyl Trifluoromethanesulfonate