missing translation for 'onlineSavingsMsg'
Learn More

o-Toluoyl chloride, 98+%, Thermo Scientific Chemicals

Catalog No. AAA1077522
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1077522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1077522 Supplier Thermo Scientific Chemicals Supplier No. A1077522

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 933-88-0 | Molecular Formula: C8H7ClO | Molecular Weight: 154.59 g/mol

o-Toluoyl chloride is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
o-Toluoyl chloride is used as a pharmaceutical intermediate.

Solubility
Reacts with water.

Notes
Keep container tightly sealed. Store under dry inert gas. Store in cool, dry conditions in well sealed containers. Incompatible with bases, active metals, alcohols,amines and oxidizing agents. It is sensitive to moisture.

Chemical Identifiers

CAS 933-88-0
Molecular Formula C8H7ClO
Molecular Weight (g/mol) 154.59
MDL Number MFCD00000668
InChI Key GPZXFICWCMCQPF-UHFFFAOYSA-N
Synonym o-toluoyl chloride, benzoyl chloride, 2-methyl, o-toluoylchloride, methylbenzoyl chloride, o-methylbenzoyl chloride, 2-toluoyl chloride, o-toluic acid chloride, unii-j9ykn18e7r, o-toluyl chloride, 2-methylbenzoic acid chloride
PubChem CID 70276
IUPAC Name 2-methylbenzoyl chloride
SMILES CC1=CC=CC=C1C(Cl)=O

Specifications

Density 1.187
Boiling Point 212°C to 214°C
Flash Point 76°C (168°F)
Odor Pungent
Refractive Index 1.5549
Quantity 100 g
UN Number UN3265
Beilstein 507933
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 154.6
Percent Purity ≥98%
Chemical Name or Material o-Toluoyl chloride
Show More Show Less
Hazard Category H227-H314
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 213-273-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only