missing translation for 'onlineSavingsMsg'
Learn More
Learn More
o-Toluenethiol 97.0+%, TCI America™
Supplier: TCI America T0288100ML
Specifications
| o-Toluenethiol | |
| Yellow | |
| C7H8S | |
| 100 mL | |
| LXUNZSDDXMPKLP-UHFFFAOYSA-N | |
| 2-methylbenzene-1-thiol | |
| 8712 | |
| ≥97.0% (GC) |
| 137-06-4 | |
| 194°C | |
| MFCD00004838 | |
| o-toluenethiol, o-thiocresol, 2-methylthiophenol, 2-thiocresol, 2-toluenethiol, o-tolyl mercaptan, o-mercaptotoluene, o-methylthiophenol, o-methylbenzenethiol, benzenethiol, 2-methyl | |
| CC1=CC=CC=C1S | |
| 124.20 | |
| 124.20 | |
| Liquid |
Chemical Identifiers
| 137-06-4 | |
| 124.20 | |
| LXUNZSDDXMPKLP-UHFFFAOYSA-N | |
| 8712 | |
| CC1=CC=CC=C1S |
| C7H8S | |
| MFCD00004838 | |
| o-toluenethiol, o-thiocresol, 2-methylthiophenol, 2-thiocresol, 2-toluenethiol, o-tolyl mercaptan, o-mercaptotoluene, o-methylthiophenol, o-methylbenzenethiol, benzenethiol, 2-methyl | |
| 2-methylbenzene-1-thiol |
Safety and Handling
RTECSNumber : XT8750000
TSCA : Yes