accessibility menu, dialog, popup

UserName

CAS RN 137-06-4

IUPAC Name: 2-methylbenzene-1-thiol
Synonyms: 2-methylbenzenethiol
Molecular Weight (g/mol): 124.2
Molecular Formula: C7H8S
InChi Key: LXUNZSDDXMPKLP-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1S
Boiling Point 
  • (2)
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (4)