missing translation for 'onlineSavingsMsg'
Learn More
Learn More
o-Toluenethiol 97.0+%, TCI America™
$417.56 - $1252.65
Chemical Identifiers
| CAS | 137-06-4 |
|---|---|
| Molecular Formula | C7H8S |
| Molecular Weight (g/mol) | 124.20 |
| MDL Number | MFCD00004838 |
| InChI Key | LXUNZSDDXMPKLP-UHFFFAOYSA-N |
| Synonym | o-toluenethiol, o-thiocresol, 2-methylthiophenol, 2-thiocresol, 2-toluenethiol, o-tolyl mercaptan, o-mercaptotoluene, o-methylthiophenol, o-methylbenzenethiol, benzenethiol, 2-methyl |
| PubChem CID | 8712 |
| IUPAC Name | 2-methylbenzene-1-thiol |
| SMILES | CC1=CC=CC=C1S |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
T028825ML
|
TCI America
T028825ML |
25 mL |
Each for $417.56
|
|
|||||
|
T0288100ML
|
TCI America
T0288100ML |
100 mL |
Each for $1,252.65
|
|
|||||
Chemical Identifiers
| 137-06-4 | |
| 124.20 | |
| LXUNZSDDXMPKLP-UHFFFAOYSA-N | |
| 8712 | |
| CC1=CC=CC=C1S |
| C7H8S | |
| MFCD00004838 | |
| o-toluenethiol, o-thiocresol, 2-methylthiophenol, 2-thiocresol, 2-toluenethiol, o-tolyl mercaptan, o-mercaptotoluene, o-methylthiophenol, o-methylbenzenethiol, benzenethiol, 2-methyl | |
| 2-methylbenzene-1-thiol |
Specifications
| 137-06-4 | |
| 194°C | |
| MFCD00004838 | |
| o-toluenethiol, o-thiocresol, 2-methylthiophenol, 2-thiocresol, 2-toluenethiol, o-tolyl mercaptan, o-mercaptotoluene, o-methylthiophenol, o-methylbenzenethiol, benzenethiol, 2-methyl | |
| CC1=CC=CC=C1S | |
| 124.20 | |
| 124.20 | |
| Liquid |
| Yellow | |
| C7H8S | |
| 25 mL | |
| LXUNZSDDXMPKLP-UHFFFAOYSA-N | |
| 2-methylbenzene-1-thiol | |
| 8712 | |
| ≥97.0% (GC) | |
| o-Toluenethiol |
Safety and Handling
RTECSNumber : XT8750000
TSCA : Yes