missing translation for 'onlineSavingsMsg'
Learn More

o-Tolidine dihydrochloride, 99%

Catalog No. AC421130250
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC421130250 25 g Glass Bottle
AC421131000 100 g Glass Bottle
AC421135000 500 g Glass Bottle
AC421130025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC421130250 Supplier Thermo Scientific Chemicals Supplier No. 421130250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 612-82-8 | C14H16N2·2HCl | 285.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 612-82-8
Molecular Formula C14H16N2·2HCl
Molecular Weight (g/mol) 285.2
InChI Key LUKPNZHXJRJBAN-UHFFFAOYSA-N
Synonym 3,3'-dimethylbenzidine dihydrochloride, o-tolidine dihydrochloride, 3,3'-dimethyl-1,1'-biphenyl-4,4'-diamine dihydrochloride, unii-5msk350kd8, dsstox_cid_511, dsstox_rid_75633, dsstox_gsid_20511, tolidine dihydrochloride, 1,1'-biphenyl-4,4'-diamine, 3,3'-dimethyl-, dihydrochloride, 3,3'-dimethylbenzidine.2hcl
PubChem CID 108938
IUPAC Name 4-(4-amino-3-methylphenyl)-2-methylaniline;dihydrochloride
SMILES CC1=C(C=CC(=C1)C2=CC(=C(C=C2)N)C)N.Cl.Cl

Specifications

Melting Point 130.0°C
Color Beige to Beige-Gray
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (on dry substance) (Argentometry)
Packaging Glass Bottle
Quantity 25 g
Merck Index 15,9675
Formula Weight 285.2
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material o-Tolidine dihydrochloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause cancer.
Harmful if swallowed.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
EINECSNumber 210-322-5

RUO – Research Use Only